Assay Details
| Gene Symbol: SLC9B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006714093.3 | XP_006714156.1 | 2 - 3 | 164 | 130 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006714093.3 transcripts.
Set Membership: |
|

SLC9B1
Genomic Map