Assay Details
| Gene Symbol: SPOCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004598.3 | NP_004589.1 | 3 - 4 | 386 | 76 | |
| GenBank mRNA | AB209701.1 | - | 4 - 5 | 370 | 76 | |
| AF231124.1 | - | 3 - 4 | 256 | 76 | ||
| AK094489.1 | - | 2 - 3 | 426 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001816.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
neuron migration
cell adhesion
signal transduction
nervous system development
negative regulation of cell-substrate adhesion
negative regulation of endopeptidase activity
negative regulation of neuron projection development
central nervous system neuron differentiation
neurogenesis
serine-type endopeptidase inhibitor activity
cysteine-type endopeptidase inhibitor activity
calcium ion binding
metalloendopeptidase inhibitor activity
regulation of cell growth
neuron migration
cell adhesion
signal transduction
nervous system development
negative regulation of cell-substrate adhesion
negative regulation of endopeptidase activity
negative regulation of neuron projection development
central nervous system neuron differentiation
neurogenesis
serine-type endopeptidase inhibitor activity
cysteine-type endopeptidase inhibitor activity
calcium ion binding
metalloendopeptidase inhibitor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05024652.hg.1 | TC05001816.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024652.hg.1 | ENST00000282223.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024652.hg.1 | ENST00000394945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024652.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000508642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000508642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000508642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000508642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000509293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000509293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183994.hg.1 | TC0500012138.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000282223.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000394945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000503916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000505690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000508642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000508642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000508642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000508642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000509293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000509293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000509978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000510689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | ENST00000515091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012138.hg.1 | NM_004598.3 | Refseq |

SPOCK1
Genomic Map