| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000231721.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000465657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000465657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000465657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000465657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000465657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000465657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000465657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000465657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000465657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000475739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000475739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000475739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000475739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000475739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000475739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000475739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
ENST00000475739.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011171.hg.1 |
NM_020163.2 |
Refseq |