Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000464177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000464177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000464177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000471255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000471255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000471255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000497754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000497754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
ENST00000497754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007242.hg.1 |
TC0X000538.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007242.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007242.hg.1 |
ENST00000372068.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007242.hg.1 |
ENST00000372072.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007242.hg.1 |
ENST00000372073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007242.hg.1 |
ENST00000464177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007242.hg.1 |
ENST00000501872.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007242.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007242.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000464177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000464177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000464177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000471255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000471255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000471255.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000497754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000497754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
ENST00000497754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008108.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000464177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000288381.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000372068.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000372072.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000372073.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000461715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
ENST00000464177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
NM_017698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
NM_032227.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155482.hg.1 |
TC0X00008108.hg.1 |
Clariom D TC |