Assay Details
Gene Symbol: NFKBIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243116.1 | NP_001230045.1 | 2 - 3 | 144 | 99 | |
NM_002503.4 | NP_002494.2 | 2 - 3 | 386 | 99 | ||
NR_040515.1 | 2 - 3 | 330 | 99 | |||
XM_006723226.2 | XP_006723289.2 | 2 - 3 | 322 | 99 | ||
XM_006723227.3 | XP_006723290.3 | 2 - 3 | 285 | 99 | ||
GenBank mRNA | AB209784.1 | - | 2 - 3 | 352 | 99 | |
AK057862.1 | - | 2 - 3 | 281 | 99 | ||
AK290569.1 | - | 2 - 3 | 337 | 99 | ||
BC015528.1 | - | 2 - 3 | 317 | 3903794 | 99 | |
BI520169.1 | - | 2 - 3 | 144 | 99 | ||
BM008311.1 | - | 2 - 3 | 338 | 99 | ||
BM449613.1 | - | 2 - 3 | 341 | 99 | ||
BQ440907.1 | - | 2 - 3 | 118 | 99 | ||
BT006743.1 | - | 2 - 3 | 283 | 99 | ||
DA633677.1 | - | 2 - 3 | 408 | 99 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000534.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001243116.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19007259.hg.1 | ENST00000313582.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007259.hg.1 | ENST00000392079.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007259.hg.1 | ENST00000509705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007259.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007259.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007259.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007259.hg.1 | TC19000534.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007252.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007252.hg.1 | TC19000534.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900160359.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160359.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160359.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160359.hg.1 | TC1900008035.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160359.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160359.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160359.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | TC1900008035.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160372.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
