Assay Details
Gene Symbol: NFKBIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243116.1 | NP_001230045.1 | 4 - 5 | 569 | 73 | |
NM_002503.4 | NP_002494.2 | 4 - 5 | 811 | 73 | ||
NR_040515.1 | 4 - 5 | 755 | 73 | |||
XM_006723226.2 | XP_006723289.2 | 4 - 5 | 747 | 73 | ||
XM_006723227.3 | XP_006723290.3 | 4 - 5 | 710 | 73 | ||
GenBank mRNA | AB209784.1 | - | 4 - 5 | 777 | 73 | |
AI684894.1 | - | 2 - 3 | 73 | |||
AK057862.1 | - | 4 - 5 | 706 | 73 | ||
AK290569.1 | - | 4 - 5 | 762 | 73 | ||
BC007197.1 | - | 2 - 3 | 411 | 73 | ||
BC015528.1 | - | 4 - 5 | 742 | 3903794 | 73 | |
BI520169.1 | - | 4 - 5 | 569 | 73 | ||
BQ440907.1 | - | 4 - 5 | 543 | 73 | ||
BT006743.1 | - | 4 - 5 | 708 | 73 | ||
BU634404.1 | - | 2 - 3 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000534.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007261.hg.1 | ENST00000313582.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007261.hg.1 | ENST00000392079.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007261.hg.1 | ENST00000509705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007261.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007261.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007261.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007261.hg.1 | TC19000534.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19007262.hg.1 | ENST00000392079.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007262.hg.1 | ENST00000509705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007262.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007262.hg.1 | TC19000534.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | TC1900008035.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160375.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160377.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160377.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160377.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160377.hg.1 | TC1900008035.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160377.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160377.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160377.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160378.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160378.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160378.hg.1 | TC1900008035.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160378.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160378.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000575359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008035.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | NR_040515.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | TC1900008035.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000313582.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000392079.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000509705.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000572515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | ENST00000576510.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | NM_001243116.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | NM_002503.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900160374.hg.1 | NR_040515.1 | Refseq |
