Assay Details
| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032352.3 | NP_115728.2 | 2 - 3 | 355 | 142 | |
| XM_005268128.1 | XP_005268185.1 | 2 - 3 | 428 | 142 | ||
| GenBank mRNA | AK310359.1 | - | 2 - 3 | 289 | 142 | |
| AK315424.1 | - | 2 - 3 | 286 | 142 | ||
| AY827074.1 | - | 2 - 3 | 341 | 142 | ||
| BC006250.2 | - | 2 - 3 | 335 | 3947694 | 142 | |
| BU507838.1 | - | 2 - 3 | 399 | 142 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000225.hg
Additional Information:
This assay is in the 5'UTR region of the AK315424.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14002438.hg.1 | ENST00000216807.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14002438.hg.1 | ENST00000543183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14002438.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14002438.hg.1 | TC14000225.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148055.hg.1 | TC1400006913.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |

BRMS1L
Genomic Map