Assay Details
| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032352.3 | NP_115728.2 | 8 - 9 | 850 | 103 | |
| XM_006720275.2 | XP_006720338.1 | 9 - 10 | 913 | 103 | ||
| XM_017021704.1 | XP_016877193.1 | 9 - 10 | 824 | 103 | ||
| XM_017021705.1 | XP_016877194.1 | 8 - 9 | 747 | 103 | ||
| GenBank mRNA | AK096496.1 | - | 7 - 8 | 573 | 103 | |
| AK297106.1 | - | 9 - 10 | 913 | 103 | ||
| AK309431.1 | - | 6 - 7 | 103 | |||
| AK315424.1 | - | 8 - 9 | 781 | 103 | ||
| AY827074.1 | - | 8 - 9 | 836 | 103 | ||
| BC006250.2 | - | 8 - 9 | 830 | 3947694 | 103 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000225.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14002444.hg.1 | ENST00000216807.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14002444.hg.1 | ENST00000543183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14002444.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14002444.hg.1 | TC14000225.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400148064.hg.1 | TC1400006913.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000216807.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000547420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000548758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000551774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552677.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | ENST00000552849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006913.hg.1 | NM_032352.3 | Refseq |

BRMS1L
Genomic Map