Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020871.hg.1 |
ENST00000257905.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020871.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020871.hg.1 |
TC12001574.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200183343.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200183343.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200183343.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200183343.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200183343.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200183343.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200183343.hg.1 |
TC1200010842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000257905.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000547826.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
ENST00000553113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010842.hg.1 |
NM_006741.3 |
Refseq |