Assay Details
| Gene Symbol: PSTPIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024430.3 | NP_077748.3 | 11 - 12 | 933 | 77 | |
| XM_011526253.2 | XP_011524555.1 | 10 - 11 | 898 | 77 | ||
| XM_011526254.1 | XP_011524556.1 | 10 - 11 | 810 | 77 | ||
| XM_011526256.1 | XP_011524558.1 | 10 - 11 | 809 | 77 | ||
| GenBank mRNA | AK023100.1 | - | 7 - 8 | 501 | 77 | |
| DA943980.1 | - | 2 - 3 | 191 | 77 | ||
| KU178456.1 | - | 11 - 12 | 837 | 77 | ||
| KU178457.1 | - | 10 - 11 | 759 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000477.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18004911.hg.1 | ENST00000360076.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004911.hg.1 | ENST00000409746.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004911.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004911.hg.1 | TC18000477.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158538.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158538.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158538.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158538.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158538.hg.1 | TC1800008580.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800158538.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158538.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158538.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158538.hg.1 | NM_024430.3 | Refseq |

PSTPIP2
Genomic Map