Assay Details
| Gene Symbol: PSTPIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024430.3 | NP_077748.3 | 6 - 7 | 509 | 136 | |
| XM_011526253.2 | XP_011524555.1 | 5 - 6 | 474 | 136 | ||
| XM_011526254.1 | XP_011524556.1 | 6 - 7 | 485 | 136 | ||
| XM_011526256.1 | XP_011524558.1 | 5 - 6 | 385 | 136 | ||
| GenBank mRNA | BC035395.1 | - | 6 - 7 | 482 | 4524856 | 136 |
| BP352927.1 | - | 6 - 7 | 509 | 136 | ||
| KU178455.1 | - | 6 - 7 | 413 | 136 | ||
| KU178456.1 | - | 6 - 7 | 413 | 136 | ||
| KU178457.1 | - | 5 - 6 | 335 | 136 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000477.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004918.hg.1 | ENST00000360076.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004918.hg.1 | ENST00000409746.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004918.hg.1 | ENST00000495213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004918.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004918.hg.1 | TC18000477.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000587042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | ENST00000593086.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008580.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | NM_024430.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | TC1800008580.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | ENST00000409746.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | ENST00000588801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | ENST00000589328.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | ENST00000591729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158545.hg.1 | NM_024430.3 | Refseq |

PSTPIP2
Genomic Map