Assay Details
Gene Symbol: CCNI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006835.2 | NP_006826.1 | 4 - 5 | 864 | 149 | |
GenBank mRNA | AB209766.1 | - | 4 - 5 | 866 | 149 | |
AF135162.1 | - | 4 - 5 | 519 | 149 | ||
AK222931.1 | - | 3 - 4 | 316 | 149 | ||
AK301111.1 | - | 3 - 4 | 725 | 149 | ||
AK312632.1 | - | 6 - 7 | 734 | 149 | ||
BC000420.2 | - | 4 - 5 | 863 | 2964432 | 149 | |
BC004975.1 | - | 4 - 5 | 863 | 2957878 | 149 | |
BT020019.1 | - | 3 - 4 | 320 | 149 | ||
CR541783.1 | - | 3 - 4 | 320 | 149 | ||
D50310.1 | - | 3 - 4 | 320 | 149 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001319.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04018045.hg.1 | TC04001319.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04018045.hg.1 | ENST00000237654.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04018045.hg.1 | ENST00000537948.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04018045.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04018048.hg.1 | TC04001319.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04018048.hg.1 | ENST00000237654.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04018048.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04018048.hg.1 | ENST00000537948.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04018048.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400175540.hg.1 | TC0400011087.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400175543.hg.1 | TC0400011087.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000237654.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000504697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000505609.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000506614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000507788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000511943.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000513774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515468.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | ENST00000515790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011087.hg.1 | NM_006835.2 | Refseq |
