| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12001844.hg.1 |
ENST00000266551.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12001844.hg.1 |
ENST00000356986.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12001844.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12001844.hg.1 |
TC12000138.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12001845.hg.1 |
ENST00000266551.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12001845.hg.1 |
ENST00000356986.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12001845.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12001845.hg.1 |
TC12000138.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147273.hg.1 |
TC1200006738.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200147270.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147270.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147270.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147270.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147270.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200147270.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200147270.hg.1 |
TC1200006738.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000266551.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000356986.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000538029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000539240.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000541957.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000543895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
ENST00000544226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006738.hg.1 |
NM_005810.3 |
Refseq |