Assay Details
Gene Symbol: NDST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022569.2 | NP_072091.1 | 10 - 11 | 2801 | 74 | |
XM_017008545.1 | XP_016864034.1 | 9 - 10 | 1799 | 74 | ||
XM_017008546.1 | XP_016864035.1 | 8 - 9 | 1105 | 74 | ||
GenBank mRNA | AB036429.1 | - | 10 - 11 | 2801 | 74 | |
AK289670.1 | - | 10 - 11 | 2779 | 74 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001488.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04021116.hg.1 | TC04001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021116.hg.1 | ENST00000264363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021116.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180070.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180070.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180070.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180070.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180070.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180070.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180070.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180070.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180070.hg.1 | TC0400011659.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
