Assay Details
Gene Symbol: NDST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022569.2 | NP_072091.1 | 1 - 2 | 423 | 119 | |
GenBank mRNA | AB036429.1 | - | 1 - 2 | 423 | 119 | |
AK289670.1 | - | 1 - 2 | 401 | 119 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001488.hg
Additional Information:
This assay is in the 5'UTR region of the AB036429.1,AK289670.1,NM_022569.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04021132.hg.1 | TC04001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021132.hg.1 | ENST00000264363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021132.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021128.hg.1 | TC04001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021128.hg.1 | ENST00000264363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021128.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021130.hg.1 | TC04001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021130.hg.1 | ENST00000264363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021130.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021129.hg.1 | TC04001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021129.hg.1 | ENST00000264363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021129.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021131.hg.1 | TC04001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021131.hg.1 | ENST00000264363.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021131.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180083.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180083.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180083.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180083.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180083.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180083.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180083.hg.1 | TC0400011659.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | ENST00000504854.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180084.hg.1 | TC0400011659.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400180085.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180085.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180085.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180085.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180085.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180085.hg.1 | ENST00000514570.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180085.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180085.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180085.hg.1 | TC0400011659.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400180082.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180082.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180082.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180082.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180082.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180082.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180082.hg.1 | TC0400011659.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400180086.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180086.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180086.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180086.hg.1 | ENST00000264363.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180086.hg.1 | ENST00000613194.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180086.hg.1 | NM_022569.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180086.hg.1 | TC0400011659.hg.1 | Clariom D TC |
