Assay Details
Gene Symbol: CSRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003476.4 | NP_003467.1 | 2 - 3 | 206 | 116 | |
GenBank mRNA | AF121260.1 | - | 1 - 2 | 96 | 116 | |
BC024010.1 | - | 2 - 3 | 209 | 4826902 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001477.hg
Additional Information:
This assay is in the 5'UTR region of the AF121260.1,BC024010.1,NM_003476.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel remodeling
regulation of the force of heart contraction
cardiac muscle hypertrophy
cellular calcium ion homeostasis
skeletal muscle tissue development
protein localization to organelle
detection of muscle stretch
positive regulation of transcription from RNA polymerase II promoter
cardiac muscle tissue development
cardiac myofibril assembly
cardiac muscle contraction
protein binding
zinc ion binding
structural constituent of muscle
telethonin binding
actinin binding
blood vessel remodeling
regulation of the force of heart contraction
cardiac muscle hypertrophy
cellular calcium ion homeostasis
skeletal muscle tissue development
protein localization to organelle
detection of muscle stretch
positive regulation of transcription from RNA polymerase II promoter
cardiac muscle tissue development
cardiac myofibril assembly
cardiac muscle contraction
protein binding
zinc ion binding
structural constituent of muscle
telethonin binding
actinin binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11018698.hg.1 | ENST00000265968.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018698.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018698.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018698.hg.1 | TC11001477.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018694.hg.1 | ENST00000265968.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018694.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018694.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018694.hg.1 | TC11001477.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11018697.hg.1 | ENST00000265968.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018697.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018697.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018697.hg.1 | TC11001477.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11018695.hg.1 | ENST00000265968.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018695.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018695.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018695.hg.1 | TC11001477.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100178302.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178302.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178302.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178302.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178302.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178302.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178302.hg.1 | TC1100010289.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100178303.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178303.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178303.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178303.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178303.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178303.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178303.hg.1 | TC1100010289.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100178304.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178304.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178304.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178304.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178304.hg.1 | TC1100010289.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010289.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178299.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178299.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178299.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178299.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178299.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178299.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178299.hg.1 | TC1100010289.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100178300.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178300.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178300.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178300.hg.1 | ENST00000265968.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178300.hg.1 | ENST00000533783.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178300.hg.1 | NM_003476.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178300.hg.1 | TC1100010289.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 117727 | - | - |
