Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018693.hg.1 |
ENST00000265968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018693.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018693.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018693.hg.1 |
TC11001477.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178298.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178298.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178298.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178298.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178298.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178298.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178298.hg.1 |
TC1100010289.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000265968.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
ENST00000533783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010289.hg.1 |
NM_003476.4 |
Refseq |