Assay Details
| Gene Symbol: ARRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020251.3 | NP_064647.1 | 12 - 13 | 1222 | 102 | |
| XM_011545035.2 | XP_011543337.1 | 13 - 14 | 1106 | 102 | ||
| XM_017017751.1 | XP_016873240.1 | 13 - 14 | 1106 | 102 | ||
| XM_017017754.1 | XP_016873243.1 | 12 - 13 | 1062 | 102 | ||
| GenBank mRNA | AF084940.1 | - | 12 - 13 | 102 | ||
| BX402200.2 | - | 3 - 4 | 102 | |||
| FJ348262.1 | - | 12 - 13 | 998 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

ARRB1
Genomic Map