Assay Details
| Gene Symbol: BORA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286746.1 | NP_001273675.1 | 4 - 5 | 671 | 227 | |
| NM_001286747.1 | NP_001273676.1 | 3 - 4 | 466 | 120 | ||
| NM_024808.3 | NP_079084.3 | 4 - 5 | 573 | 227 | ||
| GenBank mRNA | AK291496.1 | - | 4 - 5 | 399 | 227 | |
| AK298608.1 | - | 3 - 4 | 292 | 120 | ||
| BC025367.2 | - | 4 - 5 | 384 | 4098541 | 227 | |
| BC044213.1 | - | 3 - 4 | 197 | 227 | ||
| BC056876.1 | - | 4 - 5 | 396 | 6473892 | 227 | |
| BM923278.1 | - | 4 - 5 | 582 | 227 | ||
| BX648605.1 | - | 4 - 5 | 671 | 227 | ||
| EU834129.1 | - | 4 - 5 | 479 | 227 | ||
More Information
Set Membership: |
|

BORA
Genomic Map