Assay Details
| Gene Symbol: SVIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323599.1 | NP_001310528.1 | 10 - 11 | 1796 | 71 | |
| NM_021738.2 | NP_068506.2 | 9 - 10 | 2763 | 71 | ||
| XM_005252570.2 | XP_005252627.1 | 11 - 12 | 2660 | 71 | ||
| XM_005252571.3 | XP_005252628.1 | 11 - 12 | 2586 | 71 | ||
| XM_005252573.2 | XP_005252630.1 | 9 - 10 | 2792 | 71 | ||
| XM_011519630.2 | XP_011517932.2 | 9 - 10 | 3505 | 71 | ||
| XM_011519631.2 | XP_011517933.2 | 9 - 10 | 3505 | 71 | ||
| XM_011519633.2 | XP_011517935.2 | 9 - 10 | 3505 | 71 | ||
| XM_011519634.1 | XP_011517936.1 | 11 - 12 | 2718 | 71 | ||
| XM_011519635.1 | XP_011517937.1 | 9 - 10 | 2685 | 71 | ||
| XM_011519636.2 | XP_011517938.2 | 8 - 9 | 2575 | 71 | ||
| XM_017016575.1 | XP_016872064.1 | 9 - 10 | 3505 | 71 | ||
| XM_017016576.1 | XP_016872065.1 | 8 - 9 | 3424 | 71 | ||
| XM_017016577.1 | XP_016872066.1 | 9 - 10 | 3505 | 71 | ||
| XM_017016578.1 | XP_016872067.1 | 8 - 9 | 2575 | 71 | ||
| XM_017016579.1 | XP_016872068.1 | 8 - 9 | 2575 | 71 | ||
| XM_017016583.1 | XP_016872072.1 | 9 - 10 | 3505 | 71 | ||
| GenBank mRNA | AF109135.1 | - | 9 - 10 | 2763 | 71 | |
| JX467682.1 | - | 5 - 6 | 2010 | 71 | ||
More Information
Set Membership: |
|

SVIL
Genomic Map