Assay Details
| Gene Symbol: RORA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134260.2 | NP_599022.1 | 3 - 4 | 450 | 135 | |
| XM_017022466.1 | XP_016877955.1 | 1 - 2 | 2142 | 135 | ||
| GenBank mRNA | U04898.1 | - | 3 - 4 | 383 | 135 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017022466.1 transcripts.
Set Membership: |
|

RORA
Genomic Map