Assay Details
Gene Symbol: MSMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006745.4 | NP_006736.1 | 1 - 2 | 103 | 84 | |
GenBank mRNA | AK292418.1 | - | 1 - 2 | 48 | 84 | |
AK309206.1 | - | 1 - 2 | 48 | 84 | ||
BC107879.1 | - | 1 - 2 | 65 | 6498540 | 84 | |
BX441001.2 | - | 1 - 2 | 103 | 84 | ||
U93162.1 | - | 1 - 2 | 63 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000828.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK292418.1,BC107879.1,NM_006745.4,U93162.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400008465.hg.1 | ENST00000504097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008465.hg.1 | ENST00000504097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000505270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000505270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04011423.hg.1 | TC04000828.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04011423.hg.1 | ENST00000261507.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011423.hg.1 | ENST00000393766.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011423.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011423.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04011423.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04011428.hg.1 | TC04000828.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04011428.hg.1 | ENST00000261507.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011428.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04011428.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | ENST00000505270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | ENST00000505270.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163365.hg.1 | TC0400009221.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400008465.hg.1 | ENST00000504097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008465.hg.1 | ENST00000504097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000505270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000505270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009221.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | ENST00000261507.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | ENST00000393766.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | ENST00000504317.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | ENST00000507013.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | NM_001017369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | NM_006745.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400163359.hg.1 | TC0400009221.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 157577 | - | - |
