Assay Details
Gene Symbol: TRAPPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166621.1 | NP_001160093.1 | 3 - 4 | 230 | 65 | |
NM_021210.4 | NP_067033.1 | 2 - 3 | 290 | 65 | ||
GenBank mRNA | AK314038.1 | - | 2 - 3 | 296 | 65 | |
BC025413.1 | - | 2 - 3 | 149 | 65 | ||
BC032717.1 | - | 2 - 3 | 292 | 5518603 | 65 | |
BI457771.1 | - | 2 - 3 | 292 | 65 | ||
BI600633.1 | - | 3 - 4 | 235 | 65 | ||
BM762006.1 | - | 3 - 4 | 204 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001098.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17014776.hg.1 | ENST00000303731.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17014776.hg.1 | ENST00000540486.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17014776.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17014776.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17014776.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17014776.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17014776.hg.1 | TC17001098.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205172.hg.1 | TC1700012352.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000303731.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000540486.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000571739.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000571947.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000572656.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | ENST00000575639.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | NM_001166621.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | NM_021210.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | NR_030684.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | NR_030697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205173.hg.1 | TC1700012352.hg.1 | Clariom D TC |
