Assay Details
Gene Symbol: LDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002301.4 | NP_002292.1 | 1 - 2 | 151 | 70 | |
XM_017017721.1 | XP_016873210.1 | 1 - 2 | 159 | 70 | ||
GenBank mRNA | AK058181.1 | - | 1 - 2 | 73 | 70 | |
BC019249.2 | - | 1 - 2 | 104 | 4839028 | 70 | |
BC064388.1 | - | 1 - 2 | 120 | 5744600 | 70 | |
HQ386001.1 | - | 1 - 2 | 79 | 70 | ||
HQ386002.1 | - | 1 - 2 | 79 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000242.hg
Additional Information:
This assay is in the 5'UTR region of the BC019249.2,BC064388.1,HQ386001.1,NM_002301.4,XM_017017721.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003220.hg.1 | ENST00000280704.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003220.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003220.hg.1 | TC11000242.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000396215.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000535809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000536880.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000537486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000539369.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000541669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000544105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000545848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | ENST00000546146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007005.hg.1 | NM_017448.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149370.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149370.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149370.hg.1 | ENST00000280704.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149370.hg.1 | NM_002301.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149370.hg.1 | TC1100007005.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 162394 | - | - |
