Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000536474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000536474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000536474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025406.hg.1 |
ENST00000536474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025406.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025406.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025406.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025406.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025406.hg.1 |
TC12001955.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
ENST00000536474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
ENST00000536474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200192154.hg.1 |
TC1200011880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000536474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000536474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000536474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
ENST00000541723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026661.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011880.hg.1 |
NR_026662.2 |
Refseq |