Assay Details
| Gene Symbol: SLTM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006720686.3 | XP_006720749.3 | 4 - 5 | 1098 | 137 | |
| XM_011522025.1 | XP_011520327.1 | 5 - 6 | 1167 | 185 | ||
| XM_011522026.2 | XP_011520328.2 | 5 - 6 | 1146 | 185 | ||
| XM_011522027.1 | XP_011520329.1 | 4 - 5 | 1119 | 137 | ||
| XM_011522031.2 | XP_011520333.1 | 3 - 4 | 201 | 137 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011522031.2 transcripts.
Set Membership: |
|

SLTM
Genomic Map