Assay Details
| Gene Symbol: RRAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128850.1 | NP_001122322.1 | 3 - 4 | 594 | 59 | |
| NM_004165.2 | NP_004156.1 | 3 - 4 | 585 | 59 | ||
| GenBank mRNA | AK290613.1 | - | 3 - 4 | 584 | 59 | |
| BC011645.2 | - | 3 - 4 | 576 | 4299345 | 59 | |
| BC057815.1 | - | 3 - 4 | 593 | 30341066 | 59 | |
| L24564.1 | - | 3 - 4 | 565 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001177.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016358.hg.1 | ENST00000299759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016358.hg.1 | ENST00000420652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016358.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016358.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016358.hg.1 | TC16001177.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181573.hg.1 | TC1600010603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181574.hg.1 | TC1600010603.hg.1 | Clariom D TC |

RRAD
Genomic Map