| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005435.hg.1 |
ENST00000307844.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005435.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005435.hg.1 |
TC18000523.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159888.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159888.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159888.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159888.hg.1 |
TC1800008734.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800159888.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159888.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159888.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000307844.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000577499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000581310.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
ENST00000584040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008734.hg.1 |
NM_139171.1 |
Refseq |