Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
ENST00000571771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015183.hg.1 |
ENST00000330255.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015183.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015183.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015183.hg.1 |
TC17001126.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700205334.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205334.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205334.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205334.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205334.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205334.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205334.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205334.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205334.hg.1 |
TC1700012357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000330255.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
ENST00000571771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001105571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012357.hg.1 |
NM_001220493.1 |
Refseq |