Assay Details
| Gene Symbol: C17orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105520.1 | NP_001098990.1 | 1 | 1063 | 103 | |
| GenBank mRNA | BC038956.1 | - | 1 | 1084 | 103 | |
| BC082254.1 | - | 1 | 1063 | 103 | ||
| BX488537.1 | - | 1 | 103 | |||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30760989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210506 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30760952 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30760951 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210508 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16939668 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210516 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30760950 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30760954 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16939669 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30760979 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30760985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210503 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30760946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210521 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210522 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30760935 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10210513 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 924342 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P150391 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P150391 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 35591 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 49926 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P34032551 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P150391 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P150391 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16939712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16939669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16939668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760950 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760989 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760950 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16939712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16939668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16939669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210504 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10210513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30760989 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P150391 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P150391 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P34032631 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P150393 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210520 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210513 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210519 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16939667 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210522 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210509 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210504 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210506 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210503 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16939670 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210514 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210523 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16939665 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210516 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10210511 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16939721 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 229071_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11743432_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11743432_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | ENST00000624159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | ENST00000625754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | ENST00000625754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | NM_001105520.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | NM_001105520.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | NM_001105520.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006703.hg.1 | NM_001105520.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146200.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146200.hg.1 | NM_001105520.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146200.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146200.hg.1 | NM_001105520.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146200.hg.1 | TC1700006703.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | ENST00000542475.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | ENST00000624159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | ENST00000625754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | ENST00000625754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | NM_001105520.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | NM_001105520.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | NM_001105520.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006703.hg.1 | NM_001105520.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 229071_at | - | - |
| Human Genome U133 Plus 2.0 Array | 229071_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229071_at8 | - | - |
| Human Genome U95 Set | 61667_at | - | - |
| Human Genome U95 Set | 64004_PM_at | - | - |
| Human Genome U95 Set | 61667_PM_at | - | - |
| Human Genome U95 Set | 64004_at | - | - |
| Human Genome U133 Set | 229071_at | - | - |
| Human Genome U133 Set | 229071_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009014 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009014 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3237452 | CR621216.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3237452 | BC038956.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3237452 | BC052606.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3237452 | BC067087.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3237452 | BC082254.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1799031 | BG760841.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1804278 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1794987 | CR621216.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1794987 | BC038956.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1794987 | BC052606.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1794987 | BC067087.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1794987 | BC082254.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237452 | CR621216.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237452 | BC038956.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237452 | BC052606.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237452 | BC067087.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237452 | BC082254.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237452 | CR621216.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237452 | BC038956.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237452 | BC052606.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237452 | BC067087.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237452 | BC082254.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237452 | CR621216.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237452 | BC038956.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237452 | BC052606.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237452 | BC067087.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237452 | BC082254.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1799031 | BG760841.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1794987 | CR621216.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1794987 | BC038956.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1794987 | BC052606.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1794987 | BC067087.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1794987 | BC082254.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1804278 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1804278 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1799031 | BG760841.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1794987 | CR621216.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1794987 | BC038956.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1794987 | BC052606.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1794987 | BC067087.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1794987 | BC082254.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1799031 | BG760841.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1804278 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1794987 | CR621216.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1794987 | BC038956.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1794987 | BC052606.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1794987 | BC067087.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1794987 | BC082254.1 | human_transcriptome |

C17orf100
Genomic Map