Assay Details
| Gene Symbol: CASK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126054.2 | NP_001119526.1 | 11 - 12 | 1085 | 84 | |
| NM_003688.3 | NP_003679.2 | 11 - 12 | 1085 | 84 | ||
| GenBank mRNA | AB208859.1 | - | 11 - 12 | 1029 | 84 | |
| AF262404.1 | - | 6 - 7 | 525 | 84 | ||
| BC117311.1 | - | 11 - 12 | 1058 | 40125862 | 84 | |
| BC143454.1 | - | 11 - 12 | 1058 | 9051962 | 84 | |
More Information
Set Membership: |
|

CASK
Genomic Map