Assay Details
| Gene Symbol: RNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308263.1 | NP_001295192.1 | 10 - 11 | 1600 | 85 | |
| XM_017026061.1 | XP_016881550.1 | 9 - 10 | 1471 | 85 | ||
| GenBank mRNA | AB022605.1 | - | 9 - 10 | 1526 | 85 | |
| AK292561.1 | - | 10 - 11 | 1557 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

RNMT
Genomic Map