Assay Details
| Gene Symbol: SPIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123122.1 | - | 4 - 5 | 926 | 102 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000374919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000475785.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | ENST00000478405.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NM_001010862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NR_027139.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009836.hg.1 | NR_027139.1 | Refseq |

SPIN3
Genomic Map