Assay Details
Gene Symbol: TMEM164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032227.3 | NP_115603.2 | 2 - 3 | 727 | 79 | |
XM_005262205.4 | XP_005262262.1 | 2 - 3 | 753 | 79 | ||
XM_005262206.3 | XP_005262263.1 | 2 - 3 | 1022 | 79 | ||
XM_011531051.2 | XP_011529353.1 | 2 - 3 | 411 | 79 | ||
XM_017029896.1 | XP_016885385.1 | 2 - 3 | 314 | 79 | ||
XM_017029899.1 | XP_016885388.1 | 3 - 4 | 475 | 79 | ||
GenBank mRNA | BC085605.1 | - | 2 - 3 | 707 | 6142249 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000538.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011531051.2,XM_017029896.1,XM_017029899.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007236.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007236.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007236.hg.1 | ENST00000372068.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007236.hg.1 | ENST00000372073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007236.hg.1 | ENST00000501872.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007236.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007238.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007238.hg.1 | ENST00000372068.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007238.hg.1 | ENST00000372072.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007238.hg.1 | ENST00000372073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007238.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007238.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155475.hg.1 | TC0X00008108.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155477.hg.1 | TC0X00008108.hg.1 | Clariom D TC |
