Assay Details
Gene Symbol: TMEM164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017698.2 | NP_060168.2 | 2 - 3 | 181 | 67 | |
NM_032227.3 | NP_115603.2 | 3 - 4 | 772 | 67 | ||
XM_005262205.4 | XP_005262262.1 | 3 - 4 | 798 | 67 | ||
XM_005262206.3 | XP_005262263.1 | 3 - 4 | 1067 | 67 | ||
XM_005262208.4 | XP_005262265.1 | 3 - 4 | 227 | 67 | ||
XM_011531051.2 | XP_011529353.1 | 3 - 4 | 456 | 67 | ||
XM_011531054.1 | XP_011529356.1 | 3 - 4 | 445 | 67 | ||
XM_017029894.1 | XP_016885383.1 | 2 - 3 | 544 | 67 | ||
XM_017029896.1 | XP_016885385.1 | 3 - 4 | 359 | 67 | ||
XM_017029897.1 | XP_016885386.1 | 2 - 3 | 436 | 67 | ||
XM_017029899.1 | XP_016885388.1 | 4 - 5 | 520 | 67 | ||
GenBank mRNA | AK094127.1 | - | 2 - 3 | 181 | 67 | |
BC085605.1 | - | 3 - 4 | 752 | 6142249 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000538.hg
Additional Information:
This assay is in the 5'UTR region of the AK094127.1,NM_017698.2,XM_011531054.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X007239.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007239.hg.1 | ENST00000372068.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007239.hg.1 | ENST00000372072.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007239.hg.1 | ENST00000372073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007239.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007239.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007224.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007224.hg.1 | ENST00000372072.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007224.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007223.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007223.hg.1 | ENST00000372072.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007223.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155465.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155465.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155465.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155465.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155465.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155465.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155465.hg.1 | TC0X00008108.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155466.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155466.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155466.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155466.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155466.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155466.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155466.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155466.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155466.hg.1 | TC0X00008108.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155479.hg.1 | TC0X00008108.hg.1 | Clariom D TC |
