Assay Details
Gene Symbol: TMEM164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032227.3 | NP_115603.2 | 1 - 2 | 64 | 66 | |
XM_017029899.1 | XP_016885388.1 | 1 - 2 | 109 | 66 | ||
GenBank mRNA | AK026332.1 | - | 1 - 2 | 64 | 66 | |
BC085605.1 | - | 1 - 2 | 44 | 6142249 | 66 | |
BP295095.1 | - | 1 - 2 | 64 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000538.hg
Additional Information:
This assay is in the 5'UTR region of the AK026332.1,BC085605.1,NM_032227.3,XM_017029899.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X007233.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007233.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007233.hg.1 | ENST00000372068.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007233.hg.1 | ENST00000501872.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007233.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007232.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007232.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007232.hg.1 | ENST00000372068.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007232.hg.1 | ENST00000501872.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007232.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007234.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007234.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007234.hg.1 | ENST00000372068.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007234.hg.1 | ENST00000372073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007234.hg.1 | ENST00000501872.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007234.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007231.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007231.hg.1 | ENST00000372068.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007231.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007235.hg.1 | TC0X000538.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007235.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007235.hg.1 | ENST00000372068.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007235.hg.1 | ENST00000372073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007235.hg.1 | ENST00000501872.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007235.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155473.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155473.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155473.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155473.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155473.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155473.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155473.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155473.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155473.hg.1 | TC0X00008108.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372072.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000461715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000464177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000471255.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_017698.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008108.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155474.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155474.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155474.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155474.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155474.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155474.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155474.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155474.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155474.hg.1 | TC0X00008108.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | ENST00000288381.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | ENST00000372068.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | ENST00000372073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | ENST00000497754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | NM_032227.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155472.hg.1 | TC0X00008108.hg.1 | Clariom D TC |
