Assay Details
| Gene Symbol: BCAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005257472.1 | XP_005257529.1 | 25 - 26 | 2764 | 67 | |
| XM_011524939.1 | XP_011523241.1 | 28 - 29 | 3133 | 67 | ||
| XM_011524940.1 | XP_011523242.1 | 27 - 28 | 3088 | 67 | ||
| XM_011524941.1 | XP_011523243.1 | 26 - 27 | 2998 | 67 | ||
| XM_011524942.2 | XP_011523244.1 | 26 - 27 | 2854 | 67 | ||
| XM_011524943.2 | XP_011523245.1 | 25 - 26 | 2809 | 67 | ||
| XM_017024786.1 | XP_016880275.1 | 27 - 28 | 3043 | 67 | ||
| XM_017024787.1 | XP_016880276.1 | 27 - 28 | 2995 | 67 | ||
| XM_017024791.1 | XP_016880280.1 | 24 - 25 | 2719 | 67 | ||
| GenBank mRNA | AK000135.1 | - | 10 - 11 | 1246 | 67 | |
| AL831895.1 | - | 11 - 12 | 1284 | 67 | ||
More Information
Set Membership: |
|

BCAS3
Genomic Map