Assay Details
| Gene Symbol: SLC35D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007001.2 | NP_008932.2 | 9 - 10 | 825 | 107 | |
| NR_104627.1 | 9 - 10 | 825 | 107 | |||
| XM_005251674.1 | XP_005251731.1 | 9 - 10 | 800 | 107 | ||
| XM_005251675.2 | XP_005251732.1 | 7 - 8 | 669 | 107 | ||
| XM_005251678.4 | XP_005251735.1 | 6 - 7 | 464 | 107 | ||
| XM_011518164.1 | XP_011516466.1 | 7 - 8 | 639 | 107 | ||
| GenBank mRNA | AB106537.1 | - | 9 - 10 | 758 | 107 | |
| AK307890.1 | - | 9 - 10 | 805 | 107 | ||
| BC009413.1 | - | 3 - 4 | 216 | 107 | ||
| BC113579.1 | - | 10 - 11 | 778 | 8322631 | 107 | |
| BX361048.2 | - | 9 - 10 | 825 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001374.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycosaminoglycan biosynthetic process
biological_process
carbohydrate transport
keratan sulfate biosynthetic process
pyrimidine nucleotide-sugar transmembrane transport
nucleotide-sugar transmembrane transporter activity
pyrimidine nucleotide-sugar transmembrane transporter activity
glycosaminoglycan biosynthetic process
biological_process
carbohydrate transport
keratan sulfate biosynthetic process
pyrimidine nucleotide-sugar transmembrane transport
nucleotide-sugar transmembrane transporter activity
pyrimidine nucleotide-sugar transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09017133.hg.1 | TC09001374.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017133.hg.1 | ENST00000253270.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017133.hg.1 | ENST00000375259.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017133.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174223.hg.1 | TC0900010910.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000253270.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000375259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000482643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | ENST00000490599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_001286990.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NM_007001.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010910.hg.1 | NR_104627.1 | Refseq |

SLC35D2
Genomic Map