Assay Details
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052853.3 | NP_443085.2 | 4 - 5 | 1482 | 110 | |
GenBank mRNA | AF131745.1 | - | 4 - 5 | 1481 | 110 | |
AY302592.1 | - | 4 - 5 | 1482 | 110 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07003343.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07013753.hg.1 | TC07003343.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07013753.hg.1 | ENST00000072869.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07013753.hg.1 | ENST00000476491.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07013753.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | ENST00000622666.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013467.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000072869.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000473512.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000476491.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000483369.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | ENST00000498423.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | NM_052853.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700198982.hg.1 | TC0700013467.hg.1 | Clariom D TC |
