Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001261.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001261.hg.1 |
ENST00000426690.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001261.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001261.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001261.hg.1 |
TC13000113.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300146422.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146422.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146422.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146422.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146422.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146422.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146422.hg.1 |
TC1300006817.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000380099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
ENST00000487852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006817.hg.1 |
NM_004795.3 |
Refseq |