Assay Details
Gene Symbol: LRRC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282460.1 | NP_001269389.1 | 1 - 2 | 313 | 104 | |
XM_017020428.1 | XP_016875917.1 | 1 - 2 | 337 | 104 | ||
XM_017020430.1 | XP_016875919.1 | 1 - 2 | 337 | 104 | ||
GenBank mRNA | BC030276.1 | - | 1 - 2 | 308 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC030276.1, NM_001282460.1, XM_017020428.1, XM_017020430.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007085.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148124.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148124.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148124.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148124.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148124.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148124.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148124.hg.1 | TC1300007085.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300148125.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148125.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148125.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148125.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148125.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148125.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148125.hg.1 | TC1300007085.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300148126.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148126.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148126.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148126.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148126.hg.1 | ENST00000378805.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148126.hg.1 | ENST00000446175.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148126.hg.1 | ENST00000595396.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300148126.hg.1 | NM_001282460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300148126.hg.1 | TC1300007085.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 104108 | - | - |
