Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007085.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148127.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148127.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148127.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148127.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148127.hg.1 |
ENST00000378805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148127.hg.1 |
ENST00000446175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148127.hg.1 |
ENST00000595396.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148127.hg.1 |
NM_001282460.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148127.hg.1 |
TC1300007085.hg.1 |
Clariom D TC |