Assay Details
Gene Symbol: SNRPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291916.1 | NP_001278845.1 | 3 - 4 | 322 | 84 | |
NM_006938.3 | NP_008869.1 | 3 - 4 | 445 | 84 | ||
GenBank mRNA | AB451227.1 | - | 1 | 284 | 84 | |
AB451350.1 | - | 1 | 284 | 84 | ||
AJ577268.1 | - | 3 - 4 | 377 | 84 | ||
BC001721.1 | - | 3 - 4 | 406 | 3355716 | 84 | |
BC072427.1 | - | 3 - 4 | 332 | 6464946 | 84 | |
CN354758.1 | - | 3 - 4 | 258 | 84 | ||
CR542239.1 | - | 1 | 284 | 84 | ||
J03798.1 | - | 3 - 4 | 434 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000088.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal complex assembly
spliceosomal snRNP assembly
mRNA splicing, via spliceosome
positive regulation of defense response to virus by host
RNA splicing
nuclear import
mitophagy in response to mitochondrial depolarization
xenophagy
RNA binding
protein binding
poly(A) RNA binding
spliceosomal complex assembly
spliceosomal snRNP assembly
mRNA splicing, via spliceosome
positive regulation of defense response to virus by host
RNA splicing
nuclear import
mitophagy in response to mitochondrial depolarization
xenophagy
RNA binding
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000577906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000577906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000577906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001003.hg.1 | ENST00000300413.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001003.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001003.hg.1 | TC18000088.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | TC1800006834.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146829.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | TC1800006834.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146828.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146831.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146831.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146831.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146831.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146831.hg.1 | TC1800006834.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800146831.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146831.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146831.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146831.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000577906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000577906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000577906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000579618.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006834.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146830.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146830.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146830.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146830.hg.1 | NM_006938.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146830.hg.1 | TC1800006834.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800146830.hg.1 | ENST00000300413.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146830.hg.1 | ENST00000582475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146830.hg.1 | NM_001291916.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146830.hg.1 | NM_006938.3 | Refseq |
