Assay Details
| Gene Symbol: KAZALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319303.1 | NP_001306232.1 | 5 - 6 | 1922 | 91 | |
| NR_135067.1 | 4 - 5 | 1305 | 91 | |||
| XM_017016714.1 | XP_016872203.1 | 4 - 5 | 946 | 91 | ||
| GenBank mRNA | AK172864.1 | - | 5 - 6 | 1921 | 91 | |
| BG034107.1 | - | 1 - 2 | 131 | 91 | ||
| CX871488.1 | - | 4 - 5 | 596 | 91 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

KAZALD1
Genomic Map