Assay Details
| Gene Symbol: RRAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004165.2 | NP_004156.1 | 1 - 2 | 133 | 59 | |
| GenBank mRNA | AK290613.1 | - | 1 - 2 | 132 | 59 | |
| BC011645.2 | - | 1 - 2 | 124 | 4299345 | 59 | |
| BP257026.1 | - | 1 - 2 | 133 | 59 | ||
| L24564.1 | - | 1 - 2 | 113 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001177.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290613.1,BC011645.2,L24564.1,NM_004165.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016362.hg.1 | ENST00000299759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016362.hg.1 | ENST00000420652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016362.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016362.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016362.hg.1 | TC16001177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016360.hg.1 | ENST00000299759.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016360.hg.1 | ENST00000420652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016360.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016360.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016360.hg.1 | TC16001177.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600181583.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181583.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181583.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181583.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181583.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181583.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181583.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181583.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181583.hg.1 | TC1600010603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600181580.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181580.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181580.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181580.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181580.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181580.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181580.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181580.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181580.hg.1 | TC1600010603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000299759.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000420652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000566577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000567791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | ENST00000568915.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_001128850.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010603.hg.1 | NM_004165.2 | Refseq |

RRAD
Genomic Map