Assay Details
Gene Symbol: PDLIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011516.2 | NP_001011516.1 | 4 - 5 | 470 | 99 | |
XM_006714070.3 | XP_006714133.1 | 4 - 5 | 443 | 99 | ||
GenBank mRNA | AK291898.1 | - | 4 - 5 | 395 | 99 | |
BC017902.1 | - | 4 - 5 | 413 | 99 | ||
DA895978.1 | - | 4 - 5 | 470 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
