Assay Details
Gene Symbol: RAET1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011535800.2 | XP_011534102.1 | 1 | 245 | 136 | |
XM_017010831.1 | XP_016866320.1 | 1 | 245 | 136 | ||
GenBank mRNA | AK091263.1 | - | 1 | 245 | 136 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RAET1E antisense RNA 1 | RAET1E-AS1 | RAET1G-AS1 | -3.4kb | Anti-sense | Chr. 6 - 149863498 - 149919508 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC105378052 | LOC105378052 | 8.2kb | Anti-sense | Chr. 6 - 149934513 - 149936319 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011535800.2, XM_017010831.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
