Assay Details
| Gene Symbol: BACH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011160.1 | XP_016866649.1 | 2 - 3 | 529 | 60 | |
| XM_017011165.1 | XP_016866654.1 | 2 - 3 | 529 | 60 | ||
| GenBank mRNA | AK309526.1 | - | 2 - 3 | 519 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017011160.1,XM_017011165.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

BACH2
Genomic Map