Assay Details
| Gene Symbol: CDC42BPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017525.2 | NP_059995.2 | 15 - 16 | 1876 | 90 | |
| XM_011545155.2 | XP_011543457.1 | 15 - 16 | 1991 | 90 | ||
| XM_011545156.2 | XP_011543458.1 | 15 - 16 | 1971 | 90 | ||
| XM_011545157.2 | XP_011543459.1 | 15 - 16 | 1992 | 90 | ||
| XM_011545158.2 | XP_011543460.1 | 15 - 16 | 1992 | 90 | ||
| XM_011545159.2 | XP_011543461.1 | 15 - 16 | 1992 | 90 | ||
| XM_011545160.2 | XP_011543462.1 | 16 - 17 | 1943 | 90 | ||
| XM_011545161.2 | XP_011543463.1 | 15 - 16 | 1992 | 90 | ||
| XM_017017996.1 | XP_016873485.1 | 15 - 16 | 1991 | 90 | ||
| XM_017017997.1 | XP_016873486.1 | 15 - 16 | 1992 | 90 | ||
| XM_017017998.1 | XP_016873487.1 | 15 - 16 | 1992 | 90 | ||
| XM_017017999.1 | XP_016873488.1 | 5 - 6 | 575 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001914.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017017999.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022846.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022846.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11022846.hg.1 | TC11001914.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185541.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185541.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185541.hg.1 | ENST00000342711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185541.hg.1 | NM_017525.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185541.hg.1 | TC1100011188.hg.1 | Clariom D TC |

CDC42BPG
Genomic Map