Assay Details
| Gene Symbol: BACH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011536040.2 | XP_011534342.1 | 1 - 2 | 459 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011536040.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein ubiquitination
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein ubiquitination
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |

BACH2
Genomic Map